missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium bromide, 98+%

Product Code. 11329705
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329705 10 g 10g
11339705 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329705 Supplier Thermo Scientific Alfa Aesar Supplier No. L19761.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65039-08-9 | C6H11BrN2 | 191.07 g/mol

An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

Solubility
Soluble in water, dichloromethane, ethanol, acetonitrile. Insoluble in ethyl acetate, ether, and alkane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-08-9
Molecular Formula C6H11BrN2
Molecular Weight (g/mol) 191.07
MDL Number MFCD03427610
InChI Key GWQYPLXGJIXMMV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
PubChem CID 2734235
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;bromide
SMILES [Br-].CCN1C=C[N+](C)=C1

Specifications

Melting Point 70°C to 73°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,dichloromethane,ethanol,acetonitrile. Insoluble in ethyl acetate,ether,and alkane.
Formula Weight 191.07
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.