missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium chloride, 97%

Product Code. 10216923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216923 100 g Glass bottle 100g
10362873 5 g Glass bottle 5g
10545301 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10216923 Supplier Thermo Scientific Acros Supplier No. 354081000
  • ONLINE EXCLUSIVE PRICE
    154.00 €
    141.00 € / 100g
    Save 13.00 €
    8% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-09-0
Molecular Formula C6H11ClN2
Molecular Weight (g/mol) 146.62
MDL Number MFCD00074843
InChI Key BMQZYMYBQZGEEY-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180
PubChem CID 2734160
ChEBI CHEBI:61327
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCN1C=C[N+](C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Yellow
Flash Point 186°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 100 g
Viscosity 61.2 mPa.s (80°C)
Formula Weight 146.62
Percent Purity 97%
Physical Form Agglomerating Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Ethyl-3-methylimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Ethyl-3-methylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.