missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium chloride, 98+%

Product Code. 11466620
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466620 25 g 25g
11456620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466620 Supplier Thermo Scientific Alfa Aesar Supplier No. H27651.14
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol

Used in cellulose processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in cellulose processing.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-09-0
Molecular Formula C6H11ClN2
Molecular Weight (g/mol) 146.62
MDL Number MFCD00074843
InChI Key BMQZYMYBQZGEEY-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180
PubChem CID 2734160
ChEBI CHEBI:61327
SMILES [Cl-].CCN1C=C[N+](C)=C1

Specifications

Melting Point 77°C to 80°C
Flash Point 186°C (367°F)
Quantity 25 g
Sensitivity Very Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;chloride
Formula Weight 146.62
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
compliance-icons
Product Identifier
  • 1-Ethyl-3-methylimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Ethyl-3-methylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.