missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium iodide, 97%

Product Code. 15495708
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495708 5 g 5g
15405718 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495708 Supplier Thermo Scientific Alfa Aesar Supplier No. H59404.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35935-34-3 | C6H11IN2 | 238.07 g/mol

It is used as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as OLED materials.

Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.

Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35935-34-3
Molecular Formula C6H11IN2
Molecular Weight (g/mol) 238.07
MDL Number MFCD03701101
InChI Key IKQCDTXBZKMPBB-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt
PubChem CID 11075478
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;iodide
SMILES [I-].CCN1C=C[N+](C)=C1

Specifications

Color White to Yellow
Quantity 5 g
Beilstein 5160333
Sensitivity Moisture sensitive
Solubility Information Soluble in water,,dichloromethane,ethanol,acetonitrile.
Formula Weight 238.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.