missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium nitrate, 98%, Thermo Scientific™

Product Code. 15415778
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415778 250 mg 250mg
15405778 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415778 Supplier Thermo Scientific Alfa Aesar Supplier No. H59448.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143314-14-1
Molecular Formula C6H11N3O3
Molecular Weight (g/mol) 173.172
MDL Number MFCD00210007
InChI Key JDOJFSVGXRJFLL-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium nitrate, 1-ethyl-3-methylimidazolium nitrate nt, emim no3, acmc-20dtjx, dsstox_cid_29073, dsstox_rid_83292, dsstox_gsid_49217, ksc170o1h, 1-ethyl-3-methyl-1h-imidazol-3-ium nitrate
PubChem CID 16211072
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;nitrate
SMILES CCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]

Specifications

Melting Point 40°C
Quantity 250 mg
UN Number UN1479
Sensitivity Hygroscopic
Formula Weight 173.17
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.