missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperazine, 98+%, ACROS Organics™

Product Code. 10450701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10450701 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10450701 Supplier Thermo Scientific Acros Supplier No. 246070250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5308-25-8
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.895g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C6H14N2
Refractive Index 1.4680 to 1.4700 (20°C, 589nm)
MDL Number MFCD00059912
Quantity 25 g
Synonym n-ethylpiperazine, piperazine, 1-ethyl, 1-ethylpiperazin, 1-ethyl-piperazine, ethylpiperazine, n-ethyl piperazine, unii-6w2bw0v73g, 1-ethyl-piperazin, ethypiperazine, ethyl piperazine
InChI Key WGCYRFWNGRMRJA-UHFFFAOYSA-N
SMILES CCN1CCNCC1
IUPAC Name 1-ethylpiperazine
Molecular Weight (g/mol) 114.192
PubChem CID 79196
Formula Weight 114.19
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 1-Ethylpiperazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.