missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperidine, 99%, Thermo Scientific Chemicals

Product Code. 11417757
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417757 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11417757 Supplier Thermo Scientific Alfa Aesar Supplier No. A16294.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 766-09-6 | Molecular Formula: C7H15N | Molecular Weight: 113.204 g/mol

In Room temperature phosphorescence of α-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application is described. The highest yields of 5-Bromopyrimidine (37%) were obtained by using 1-ethylpiperidine or triethylamine as condensing agent. Tributyltin hydride and 1-ethylpiperidine hypophosphite were mediated intermolecular radical additions to 2, 4, 6-trichlorophenyl vinyl sulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In Room temperature phosphorescence of α-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application is described. The highest yields of 5-Bromopyrimidine (37%) were obtained by using 1-ethylpiperidine or triethylamine as condensing agent. Tributyltin hydride and 1-ethylpiperidine hypophosphite were mediated intermolecular radical additions to 2, 4, 6-trichlorophenyl vinyl sulfonate.

Solubility
1-ethylpiperidine salt of benzylpenicillin is considerably less soluble in amyl acetate and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-09-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00006507
InChI Key HTLZVHNRZJPSMI-UHFFFAOYSA-N
Synonym n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene
PubChem CID 13007
ChEBI CHEBI:39017
IUPAC Name 1-ethylpiperidine
SMILES CCN1CCCCC1

Specifications

Melting Point -20°C
Density 0.825
Boiling Point 129°C to 131°C
Flash Point 18°C (64°F)
Refractive Index 1.444
Quantity 500 g
UN Number UN2386
Beilstein 102643
Solubility Information 1-ethylpiperidine salt of benzylpenicillin is considerably less soluble in amyl acetate and acetone.
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material 1-Ethylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.