missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-nitrobenzene, 99%

Product Code. 10031040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031040 50 mL Glass bottle 50mL
10112820 10 mL Glass bottle 10mL
10433775 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031040 Supplier Thermo Scientific Acros Supplier No. 119480500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
MDL Number MFCD00007048
InChI Key PWKNBLFSJAVFAB-UHFFFAOYSA-N
Synonym 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
PubChem CID 73895
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -9°C to 6°C
Color Brown to Yellow
Density 1.3350g/mL
Boiling Point 215°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NO2
Refractive Index 1.5307 to 1.5327
Quantity 50 mL
Beilstein 05, 241
Specific Gravity 1.335
Solubility Information Solubility in water: immiscible
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.