missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoronaphthalene, 98%

Product Code. 11438307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438307 100 g 100g
11418307 5 g 5g
11428307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438307 Supplier Thermo Scientific Alfa Aesar Supplier No. A18378.22
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 321-38-0 | C10H7F | 146.164 g/mol

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-38-0
Molecular Formula C10H7F
Molecular Weight (g/mol) 146.164
MDL Number MFCD00003873
InChI Key CWLKTJOTWITYSI-UHFFFAOYSA-N
Synonym fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
PubChem CID 9450
IUPAC Name 1-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2F

Specifications

Melting Point -10°C
Density 1.331
Boiling Point 215°C to 217°C
Flash Point 65°C (149°F)
Refractive Index 1.593
Quantity 100 g
Beilstein 1906413
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 146.17
Percent Purity 98%
Chemical Name or Material 1-Fluoronaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.