missing translation for 'onlineSavingsMsg'
Learn More

1-Formylpiperidine, 99%

Product Code. 11465437
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465437 250 g 250g
11455437 50 g 50g
11445437 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465437 Supplier Thermo Scientific Alfa Aesar Supplier No. B23452.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2591-86-8 | C6H11NO | 113.16 g/mol

1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.

Solubility
Freely soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2591-86-8
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006483
InChI Key FEWLNYSYJNLUOO-UHFFFAOYSA-N
Synonym n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
PubChem CID 17429
ChEBI CHEBI:42546
IUPAC Name piperidine-1-carbaldehyde
SMILES C1CCN(CC1)C=O

Specifications

Melting Point -31°C
Density 1.019
Boiling Point 222°C
Flash Point 102°C (215°F)
Refractive Index 1.484
Quantity 250 g
Beilstein 107697
Solubility Information Freely soluble in water.
Formula Weight 113.16
Percent Purity 99%
Chemical Name or Material 1-Formylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.