missing translation for 'onlineSavingsMsg'
Learn More

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

Product Code. 10605302 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
100g
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10605302 100 g Plastic powder jar 100g
10315850 25 g Plastic powder jar 25g
11483743 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10605302 Supplier Fisher Chemical Supplier No. H/0168/48
  • ONLINE EXCLUSIVE PRICE
    662.95 €
    454.00 € / 100g
    Save 208.95 €
    31% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical, Quantity: 100g, Packaging: Plastic powder jar, CAS: 22767-50-6, White, Melting Point: 300 deg.C, Molecular Formula: C7H15NaO3S, Molecular Weight: 202.244

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number 7543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
Physical Form Powder
Quantity 100 g
Formula Weight 202.25g/mol
Grade Chromatography
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • 1-Heptanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.