missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™

Product Code. 15547984 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15547984 25 g Wide Mouth Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547984 Supplier Avantor J.T.Baker Supplier No. 217305
  • ONLINE EXCLUSIVE PRICE
    258.80 €
    225.00 € / 25g
    Save 33.80 €
    13% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Specific Gravity 1L = 0.00kg
Formula Weight 202.26
Grade HPLC
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.