missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 99%

Product Code. 11495143
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495143 500 mL 500mL
11485143 100 mL 100mL
11405153 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495143 Supplier Thermo Scientific Alfa Aesar Supplier No. A12793.AP
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-70-6 | C7H16O | 116.20 g/mol

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Density 0.820
Assay Percent Range 99%
Linear Formula CH3(CH2)6OH
Quantity 500 mL
Beilstein 1731686
Merck Index 14,4661
Formula Weight 116.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptanol
compliance-icons
Product Identifier
  • 1-Heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Heptanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.