missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, TRC

Product Code. 30406578
Change view
Click to view available options
Quantity:
5 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406578 250 mL
30396632 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406578 Supplier Toronto Research Chemicals Supplier No. TRCH281195250ML
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7 H16 O
Molecular Weight (g/mol) 116.2
Synonym 1-Heptanol (9CI, ACI), Heptyl alcohol (8CI), 1-Hydroxyheptane, Enanthic alcohol, Gentanol, n-Heptan-1-ol, n-Heptanol, n-Heptyl alcohol, NSC 3703
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3
Formula Weight 116.1201
Chemical Name or Material 1-Heptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.