missing translation for 'onlineSavingsMsg'
Learn More

1-Heptene, 98+%

Product Code. 11392447
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392447 100 mL 100mL
11382447 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392447 Supplier Thermo Scientific Alfa Aesar Supplier No. L11487.AE
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 592-76-7 | C7H14 | 98.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-76-7
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009531
InChI Key ZGEGCLOFRBLKSE-UHFFFAOYSA-N
Synonym 1-heptene, heptene, heptylene, n-hept-1-ene, 1-n-heptene, unii-o748kj11v7, 1-heptene, analytical standard, n-heptene, .alpha.-heptylene, heptene petroleum
PubChem CID 11610
IUPAC Name hept-1-ene
SMILES CCCCCC=C

Specifications

Melting Point ∼-119°C
Density 0.697
Boiling Point 93°C to 94°C
Flash Point −8°C (17°F)
Linear Formula CH3(CH2)4CH=CH2
Refractive Index 1.4
Quantity 100 mL
UN Number UN2278
Beilstein 1098332
Formula Weight 98.19
Percent Purity ≥98%
Chemical Name or Material 1-Heptene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.