missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol, 96%

Product Code. 10486920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10486920 1 kg 1kg
10142820 2.5 kg 2.5kg
11994601 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10486920 Supplier Thermo Scientific Acros Supplier No. 120480010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 36653-82-4 | C16H34O | 242.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 47°C to 50°C
Color White
Boiling Point 344°C
Flash Point >157°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)15OH
Quantity 1 kg
Beilstein 01,429
Merck Index 15,2027
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethanol,soluble in acetone,very soluble in ether,benzene,chloroform
Viscosity 8 mPa.s (60°C)
Formula Weight 242.44
Percent Purity 96%
Physical Form Powder, Flakes or Pellets
Chemical Name or Material 1-Hexadecanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.