missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol, TRC

Product Code. 17909984
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909984 25 g 25g
17998774 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17909984 Supplier Toronto Research Chemicals Supplier No. TRCH29381025G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16 H34 O
Molecular Weight (g/mol) 242.44
Synonym 1-Hexadecanol (9CI, ACI), 1-Cetanol, 1-Hydroxyhexadecane, Adol 52, Adol 52NF, Adol 54, Alfol 16, Alfol 16RD, Atalco C, Cachalot C 51, Cetaffine, Cetal, Cetalcos, Cetalol CA, Cetanol, Cetanol 70, Cetanol NX, Cetyl alcohol, Cetyl Alcohol NX, Cetylic alcohol, Cetylol, CO 1695, CO 1695F, Conol 1670, Conol 1695, Crodacol C, Crodacol C 90, Crodacol C 95, Crodacol CAS, Crodacol CAT, Elfacos C, Epal 16, Ethal, Ethol, Ginol 16, Hainol 16SS, Hexadecanol, Hexadecyl alcohol, Hyfatol 16, Hyfatol 16-85, Hyfatol 16-95, Kalcohl 60, Kalcohl 6098, Kalcol 6028, Kalcol 6098, Kolliwax CA, Lanette 16, Lanette 1665, Lanol C, Laurex 16, Lipocol C, Lorol 24, Lorol C 14/98, Lorol C 16, Loxanol K, Loxanol K extra, Loxanwax SK, n-1-Hexadecanol, n-Cetyl alcohol, n-Hexadecanol, n-Hexadecyl alcohol, NAA 44, Nacol 16-95, Nacol 16-99, Nacol 1618, Nafol 16, NSC 4194, Palmitic alcohol, Palmityl alcohol, Product 308, Siponol CC, Siponol Wax A, SP Crodacol C 90MBAL, Spermol, Speziol C 16, Tego Alkanol 16
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3
Formula Weight 242.261
Percent Purity >95%
Chemical Name or Material Cetyl Alcohol (1-Hexadecanol)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.