missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene, 92%, tech.

Product Code. 10733861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10733861 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10733861 Supplier Thermo Scientific Acros Supplier No. 120500010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 629-73-2 | C16H32 | 224.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 629-73-2
CAS Min % 90.0
CAS Max % 100.0
Melting Point 3°C to 5°C
Color Brown-Orange to Yellow
Density 0.7830g/mL
Boiling Point 274°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Molecular Formula C16H32
Linear Formula CH3(CH2)13CH=CH2
Refractive Index 1.4390 to 1.443
MDL Number MFCD00008991
Quantity 1 L
Beilstein 01, 226
Specific Gravity 0.783
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,petroleum and coal-tar,solvents,soluble in hydrocarbon solvents
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCC=C
IUPAC Name hexadec-1-ene
Molecular Weight (g/mol) 224.43
PubChem CID 12395
ChEBI CHEBI:77507
Viscosity 3.83 mm2/s (20°C)
Formula Weight 224.43
Percent Purity 92%
Grade Technical
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Hexadecene, tech., 92%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hexadecene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.