missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecene, 90+%

Product Code. 11347775
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347775 250 mL 250mL
11357775 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347775 Supplier Thermo Scientific Alfa Aesar Supplier No. L02637.AK
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 629-73-2 | C16H32 | 224.432 g/mol

1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

Solubility
Miscible with ether, petroleum ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.432
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

Melting Point 4°C
Density 0.781
Boiling Point 273°C to 274°C
Flash Point 132°C (269°F)
Refractive Index 1.442
Quantity 250 mL
Beilstein 1753401
Solubility Information Miscible with ether,petroleum ether and alcohol. Immiscible with water.
Formula Weight 224.43
Percent Purity 94%
Chemical Name or Material 1-Hexadecene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.