missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)pyridinium chloride monohydrate, 98%

Product Code. 11498740
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498740 25 g 25g
11495593 100 g 100g
11405603 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498740 Supplier Thermo Scientific Alfa Aesar Supplier No. A13499.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol

(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

Solubility
Soluble in water, alcohol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

Melting Point 83°C to 86°C
Quantity 25 g
UN Number UN2811
Beilstein 3578606
Merck Index 14,2032
Solubility Information Soluble in water, alcohol, chloroform
IUPAC Name 1-hexadecylpyridin-1-ium;chloride;hydrate
Formula Weight 358.02 (339.99 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1-Hexadecyl)pyridinium chloride monohydrate
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • (1-Hexadecyl)pyridinium chloride monohydrate
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: (1-Hexadecyl)pyridinium chloride monohydrate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.