missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine

Product Code. 11450627
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
This item is not returnable. View return policy

Product Code. 11450627

Brand: Thermo Scientific Alfa Aesar B22459.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143-27-1 | C16H35N | 241.46 g/mol

1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-27-1
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00008158
InChI Key FJLUATLTXUNBOT-UHFFFAOYSA-N
Synonym hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
PubChem CID 8926
IUPAC Name hexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN

Specifications

Melting Point 42°C to 47°C
Density 0.813
Boiling Point 144°C (2 mm)
Flash Point 140°C (284°F)
Assay Percent Range 90%
Refractive Index 1.4496
Quantity 1000 g
UN Number UN3259
Beilstein 1634065
Solubility Information Insoluble in water.
Formula Weight 241.46
Percent Purity 90%
Grade Technical
Assay remainder mainly 1-octadecylamine
Chemical Name or Material 1-Hexadecylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.