missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade

Product Code. 10075670 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
Product Code. Quantity unitSize
10075670 5 g 5g
10495841 25 g 25g
10628321 100 g 100g
11350996 500 g 500g
4 options
This item is not returnable. View return policy

Product Code. 10075670

Brand: Thermo Scientific 411600050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium;hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Absorbance (0.25M aq. soln.) (1cm cell vs HPLC grade water), 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm
Melting Point >300°C
Color Colorless to Yellow
Density Negligible
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)5SO3Na·xH2O
Quantity 5 g
Beilstein 04, IV, 54
Solubility Information (10% in water) Clear colorless
Formula Weight 188.22
Vapor Pressure Negligible
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Hexanesulfonic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.