missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
Specifications
Specifications
| Absorbance | (0.25M aq. soln.) (1cm cell vs HPLC grade water), 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm |
| Melting Point | >300.0°C |
| Color | Colorless to Yellow |
| Density | Negligible |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)5SO3Na·xH2O |
| Quantity | 500 g |
| Beilstein | 04, IV, 54 |
| Show More |
Safety and Handling
Product Identifier
- 1-Hexanesulfonic acid, sodium salt hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Hexanesulfonic acid, sodium salt hydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction