missing translation for 'onlineSavingsMsg'
Learn More

1-hexanesulfonic acid, sodium salt hydrate, 99%

Product Code. 10553951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553951 25 g Glass bottle 25g
10062740 5 g Glass bottle 5g
10329233 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10553951 Supplier Thermo Scientific Acros Supplier No. 206120250
  • ONLINE EXCLUSIVE PRICE
    251.00 €
    243.00 € / 25g
    Save 8.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium;hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Absorbance 5.5 w/v% aq. soln., pH 3, H3PO4, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm, 5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm
Melting Point >300°C
Color White
Density Negligible
Loss on Drying 9.6% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5SO3Na·xH2O
Quantity 25 g
Solubility Information (10% in water) Clear colorless
Formula Weight 188.22
Vapor Pressure Negligible
Percent Purity 99%
Physical Form Crystalline Flakes or Solid
Chemical Name or Material 1-Hexanesulfonic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.