missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanethiol, 96%

Product Code. 11362187
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362187 100 mL 100mL
11352187 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362187 Supplier Thermo Scientific Alfa Aesar Supplier No. L09659.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 111-31-9 | C6H14S | 118.24 g/mol

It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-31-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00004909
InChI Key PMBXCGGQNSVESQ-UHFFFAOYSA-N
Synonym 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
PubChem CID 8106
IUPAC Name hexane-1-thiol
SMILES CCCCCCS

Specifications

Melting Point -81°C
Density 0.838
Boiling Point 150°C to 154°C
Flash Point 30°C (86°F)
Odor Strong
Linear Formula CH3(CH2)5SH
Refractive Index 1.448
Quantity 100 mL
UN Number UN3071
Beilstein 1731295
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 118.24
Percent Purity 97%
Chemical Name or Material 1-Hexanethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Hexanethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 1-Hexanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.