missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, 98%, Reagent Grade, Solstice

Product Code. 15697890
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15697890 2.5 L Glass bottle 2.5 L
15677890 100 mL Glass bottle 100mL
15687890 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15697890 Supplier Fluka Supplier No. H133032.5L
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    100.23 € / 2.5 L
    Save 21.77 €
    17% Off
 Restricted Item
Estimated Shipment: 01-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -52°C (literature)
Color Colorless
Density 0.814g/mL at 25°C
Boiling Point 156°C to 157°C (literature)
Packaging Glass bottle
Linear Formula CH3(CH2)5OH
Quantity 2.5 L
UN Number UN2282
Beilstein 969167
Formula Weight 102.17g/mol
Vapor Pressure 1mmHg (25.6°C)
Percent Purity 0.98
Chemical Name or Material 1-Hexanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Hexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: hexan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.