missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol

Product Code. 11382007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382007 25 g 25g
11372007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382007 Supplier Thermo Scientific Alfa Aesar Supplier No. L07660.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1629-60-3 | C6H10O | 98.145 g/mol

1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water (slightly).

Notes
Store at 4°C. Protect from heat. Incompatible with oxidizing agents, heat, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1629-60-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00051563
InChI Key JTHNLKXLWOXOQK-UHFFFAOYSA-N
Synonym 1-hexen-3-one, propyl vinyl ketone, unii-qh9d98z86n, 1-hexenone-3, n-propylacrolein, vinyl propyl ketone, acmc-1bt5f, 1-hexene-3-one
PubChem CID 15395
IUPAC Name hex-1-en-3-one
SMILES CCCC(=O)C=C

Specifications

Density 0.84
Boiling Point 127°C to 129°C
Flash Point 22°C (71°F)
Odor Pungent
Refractive Index 1.4275
Quantity 25 g
UN Number UN1224
Beilstein 1740796
Solubility Information Soluble in water (slightly).
Formula Weight 98.15
Percent Purity ≥90%
Chemical Name or Material 1-Hexen-3-one, stabilized with 0.5% 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.