missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene, 97%

Product Code. 10104890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Unit Size:
1 L
100mL
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10104890 100 mL Glass bottle 100mL
10348540 1 L Glass bottle 1 L
10679084 2.5 L Glass bottle 2.5 L
10689084 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10104890 Supplier Thermo Scientific Acros Supplier No. 213211000
  • ONLINE EXCLUSIVE PRICE
    27.75 €
    26.90 € / 100mL
    Save 0.85 €
    3% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15 | CAS: 592-41-6 | C6H12 | 84.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-41-6
Molecular Formula C6H12
Molecular Weight (g/mol) 84.15
MDL Number MFCD00009505
InChI Key LIKMAJRDDDTEIG-UHFFFAOYSA-N
Synonym 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
PubChem CID 11597
ChEBI CHEBI:24579
IUPAC Name hex-1-ene
SMILES CCCCC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -140.0°C
Color Colorless
Density 0.6780g/mL
Boiling Point 62.0°C to 65.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH=CH2
Refractive Index 1.3867 to 1.3887
Quantity 100 mL
Beilstein 01, 215
Specific Gravity 0.678
Solubility Information Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
Viscosity 0.34 mm2/s (40°C)
Formula Weight 84.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Hexene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 1-Hexene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.