missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl isothiocyanate, 97%

Product Code. 11409000
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409000 5 g 5g
11419000 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409000 Supplier Thermo Scientific Alfa Aesar Supplier No. A12223.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4404-45-9 | C7H13NS | 143.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4404-45-9
Molecular Formula C7H13NS
Molecular Weight (g/mol) 143.248
MDL Number MFCD00014445
InChI Key WXYAXKKXIGHXDS-UHFFFAOYSA-N
Synonym hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
PubChem CID 78120
IUPAC Name 1-isothiocyanatohexane
SMILES CCCCCCN=C=S

Specifications

Density 0.933
Boiling Point 216°C
Flash Point 90°C (194°F)
Odor Pungent
Refractive Index 1.4915
Quantity 5 g
UN Number UN2922
Beilstein 1751837
Sensitivity Moisture sensitive
Formula Weight 143.25
Percent Purity 97%
Chemical Name or Material 1-Hexyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.