missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexyl)trimethylammonium bromide, 98%

Product Code. 11342596
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342596 25 g 25g
11332596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342596 Supplier Thermo Scientific Alfa Aesar Supplier No. L08594.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2650-53-5 | C9H22BrN | 224.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2650-53-5
Molecular Formula C9H22BrN
Molecular Weight (g/mol) 224.186
MDL Number MFCD00051872
InChI Key JYVPKRHOTGQJSE-UHFFFAOYSA-M
Synonym hexyltrimethylammonium bromide, hexyltrimethylazanium bromide, acmc-209grw, hexyltrimethylammoniumbromide, n-hexyltrimethylammonium bromide, n,n,n,n-hexyltrimethylammonium bromide, 1-hexanaminium,n,n-trimethyl-, bromide, 1-hexyl trimethylammonium bromide, 1-hexanaminium,n,n,n-trimethyl-, bromide 1:1
PubChem CID 10059492
IUPAC Name hexyl(trimethyl)azanium;bromide
SMILES CCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 177°C to 180°C
Quantity 25 g
Beilstein 3913229
Sensitivity Hygroscopic
Formula Weight 224.18
Percent Purity 98%
Chemical Name or Material (1-Hexyl)trimethylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.