missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylboronic acid, 97%

Product Code. 11400607
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400607 2.5 g 2.5g
11410607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400607 Supplier Thermo Scientific Alfa Aesar Supplier No. B22446.88
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16343-08-1 | C6H15BO2 | 129.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16343-08-1
Molecular Formula C6H15BO2
Molecular Weight (g/mol) 129.994
MDL Number MFCD01074641
InChI Key CXSYDLCMCLCOCA-UHFFFAOYSA-N
Synonym n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2
PubChem CID 351064
IUPAC Name hexylboronic acid
SMILES B(CCCCCC)(O)O

Specifications

Melting Point 85°C to 90°C
Quantity 2.5 g
Beilstein 1736080
Sensitivity Air Sensitive
Formula Weight 130
Percent Purity 97%
Chemical Name or Material 1-Hexylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.