missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxycyclohexyl phenyl ketone, 98%

Product Code. 15468939
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468939 500 g 500g
15458939 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468939 Supplier Thermo Scientific Alfa Aesar Supplier No. L11377.36
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 947-19-3 | C13H16O2 | 204.27 g/mol

1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.

Solubility
Slightly soluble in water (1108 mg/L at 25°C). Soluble in acetone, butyl acetate, methanol and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-19-3
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.27
MDL Number MFCD00059561
InChI Key QNODIIQQMGDSEF-UHFFFAOYSA-N
Synonym 1-hydroxycyclohexyl phenyl ketone, 1-hydroxycyclohexyl phenyl methanone, 1-benzoylcyclohexanol, hydroxycyclohexyl phenyl ketone, 1-benzoylcyclohexan-1-ol
PubChem CID 70355
SMILES OC1(CCCCC1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 46°C to 49°C
Density 1.17
Boiling Point >225°C
Flash Point >150°C
Quantity 500 g
Beilstein 2330973
Solubility Information Slightly soluble aq. soln. (1108 mg/L (at 25°C). Soluble in acetone,butyl acetate,methanol and toluene.)
IUPAC Name 1-benzoylcyclohexan-1-ol
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material 1-Hydroxycyclohexyl phenyl ketone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.