missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxymethyl-5,5-dimethylhydantoin, 97%, Thermo Scientific™

Product Code. 11498697
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11498697 25 g 25g
11408707 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11498697

Brand: Thermo Scientific Alfa Aesar A19868.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Hydroxymethyl-5,5-dimethylhydantoin is used as chelator for light-sensitive pharmaceutical drug, and as raw material of water-soluble polymers. It also can be used as raw material of disinfectants and preservatives, and soluble inhibitor for proteins. It is used as an adjuvant in the bleaching of food-contact recycled paper and board used in food packaging.

Solubility
Freely soluble in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-25-6
Molecular Formula C6H10N2O3
Molecular Weight (g/mol) 158.16
MDL Number MFCD00020866
InChI Key SIQZJFKTROUNPI-UHFFFAOYSA-N
Synonym 1-hydroxymethyl-5,5-dimethylhydantoin, mdm hydantoin, glycoserve, mdmh, 1-hydroxymethyl-5,5-dimethylimidazolidine-2,4-dione, mdm hydantoin van, monomethyloldimethylhydantoin, 1-monomethylol-5,5-dimethylhydantoin, methylol dimethylhydantoin
PubChem CID 67000
IUPAC Name 1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)N(CO)C(=O)NC1=O

Specifications

Melting Point 104°C to 106°C
Quantity 25 g
Beilstein 139160
Merck Index 14,4831
Solubility Information Freely soluble in water.
Formula Weight 158.16
Percent Purity 97%
Chemical Name or Material 1-Hydroxymethyl-5,5-dimethylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.