missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%

Product Code. 15465045
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465045 2 g 2g
15455045 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465045 Supplier Thermo Scientific Alfa Aesar Supplier No. 046884.04
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104439-77-2 | C62H42O6Ru2

1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydroteracarbonyldiruthenium(II) is widely used in various types of applications in organic synthesis. It is used for hydrogenation of carbonyls, alkenes and imines. It is also used in the alkylation of amines and amination of propargyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104439-77-2
Molecular Formula C62H42O6Ru2
MDL Number MFCD03840556
Synonym Shvo's Catalyst

Specifications

Melting Point 223°C to 227°C
Color Yellow to Orange
Quantity 2 g
Sensitivity Moisture sensitive
Merck Index 14,8483
Solubility Information Soluble in water and polar organic solvents.
Formula Weight 1085.15
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldir

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.