missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-methylpropane, 97%, stab. with copper

Product Code. 11462598
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
1000g
100g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462598 1 kg 1000g
11494160 25 g 25g
11404170 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462598 Supplier Thermo Scientific Alfa Aesar Supplier No. A15883.0B
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 513-38-2 | C4H9I | 184.02 g/mol

1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-38-2
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001084
InChI Key BTUGGGLMQBJCBN-UHFFFAOYSA-N
Synonym isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
PubChem CID 10556
IUPAC Name 1-iodo-2-methylpropane
SMILES CC(C)CI

Specifications

Melting Point -93°C
Density 1.599
Boiling Point 120°C to 121°C
Flash Point 12°C
Refractive Index 1.496
Quantity 1 kg
UN Number UN2391
Beilstein 1730927
Sensitivity Light sensitive
Merck Index 14,5144
Solubility Information Insoluble aq. soln..
Formula Weight 184.02
Percent Purity 98%
Chemical Name or Material 1-Iodo-2-methylpropane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.