missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-methylpropane, 97%, stabilized

Product Code. 10583771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10583771 100 mL Glass bottle 100mL
10124660 25 mL Glass bottle 25mL
10063383 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10583771 Supplier Thermo Scientific Acros Supplier No. 223741000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 513-38-2 | C4H9I | 184.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-38-2
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001084
InChI Key BTUGGGLMQBJCBN-UHFFFAOYSA-N
Synonym isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
PubChem CID 10556
IUPAC Name 1-iodo-2-methylpropane
SMILES CC(C)CI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -93.0°C
Color Colorless to Orange
Density 1.5900g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2I
Refractive Index 1.4948 to 1.4968
Quantity 100 mL
Merck Index 15,5189
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 184.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-methylpropane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodo-2-methylpropane, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.