missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-methylbutane, 97%, stab. with copper

Product Code. 11371847
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371847 50 g 50g
11361847 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371847 Supplier Thermo Scientific Alfa Aesar Supplier No. L05194.18
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-28-6 | C5H11I | 198.047 g/mol

1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.

Solubility
Insoluble in water. Soluble in most common organic solvents.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-28-6
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.047
MDL Number MFCD00039414
InChI Key BUZZUHJODKQYTF-UHFFFAOYSA-N
Synonym isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butane
PubChem CID 10924
IUPAC Name 1-iodo-3-methylbutane
SMILES CC(C)CCI

Specifications

Density 1.515
Boiling Point 146°C to 148°C
Flash Point 40°C (104°F)
Refractive Index 1.492
Quantity 50 g
UN Number UN1993
Beilstein 1730990
Sensitivity Light sensitive
Merck Index 14,5120
Solubility Information Insoluble in water. Soluble in most common organic solvents.
Formula Weight 198.05
Percent Purity 97%
Chemical Name or Material 1-Iodo-3-methylbutane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.