missing translation for 'onlineSavingsMsg'
Learn More

1-Iodododecane, 98%, stabilized

Product Code. 13498488 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13498488 500 mL Glass Bottle 500mL
10247300 25 mL Glass Bottle 25mL
10075663 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13498488 Supplier Thermo Scientific Acros Supplier No. 250095000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4292-19-7 | C12H25I | 296.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4292-19-7
Molecular Formula C12H25I
Molecular Weight (g/mol) 296.24
MDL Number MFCD00001088
InChI Key GCDPERPXPREHJF-UHFFFAOYSA-N
Synonym n-dodecyl iodide, dodecyl iodide, lauryl iodide, dodecane, 1-iodo, dodecane,1-iodo, iodododecane, iodo-dodecane, 1-iodanyldodecane, pubchem3943, n-iodododecane
PubChem CID 20282
IUPAC Name 1-iodododecane
SMILES CCCCCCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 159.0°C to 160.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)11I
Refractive Index 1.4830 to 1.4850
Quantity 500 mL
Beilstein 01,67
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 296.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodododecane, Stabilized with 500ppm copper
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodododecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.