missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexadecane, 98%, stab. with copper

Product Code. 11399216
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399216 100 g 100g
15423067 25 g 25g
11309226 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399216 Supplier Thermo Scientific Alfa Aesar Supplier No. A12977.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 544-77-4 | C16H33I | 352.344 g/mol

1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodohexadecane, is used as an ionic liquid for electrochemical applications. It is also used in detection methods as primer and probe design instrumentation applications guide in qPCR Technic.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from strong oxidizing agents, strong bases. Store it at a temperature of 15 - 30°C. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-77-4
Molecular Formula C16H33I
Molecular Weight (g/mol) 352.344
MDL Number MFCD00001089
InChI Key KMWHQYDMBYABKL-UHFFFAOYSA-N
Synonym cetyl iodide, hexadecyl iodide, hexadecane, 1-iodo, hexadecane, iodo, unii-jo95x48m8o, 1-iodo-hexadecan, iodohexadecane, cetyljodid, 1-iodo-hexadecane, n-hexadecyl iodide
PubChem CID 11007
IUPAC Name 1-iodohexadecane
SMILES CCCCCCCCCCCCCCCCI

Specifications

Melting Point 21°C to 23°C
Density 1.121
Boiling Point 206°C to 207°C (10 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)15I
Refractive Index 1.482
Quantity 100 g
Beilstein 1748533
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 352.34
Percent Purity 98%
Chemical Name or Material 1-Iodohexadecane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.