missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexane, 98%, stab. with copper

Product Code. 11404854
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404854 100 g 100g
15453077 25 g 25g
11414854 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404854 Supplier Thermo Scientific Alfa Aesar Supplier No. A13784.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 638-45-9 | C6H13I | 212.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-45-9
Molecular Formula C6H13I
Molecular Weight (g/mol) 212.074
MDL Number MFCD00001102
InChI Key ANOOTOPTCJRUPK-UHFFFAOYSA-N
Synonym hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
PubChem CID 12527
IUPAC Name 1-iodohexane
SMILES CCCCCCI

Specifications

Melting Point -75°C
Density 1.437
Boiling Point 179°C to 181°C
Flash Point 61°C (141°F)
Refractive Index 1.492
Quantity 100 g
Beilstein 505971
Sensitivity Light sensitive
Formula Weight 212.08
Percent Purity 98%
Chemical Name or Material 1-Iodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.