missing translation for 'onlineSavingsMsg'
Learn More

1-Iodonaphthalene, 98%

Product Code. 11445257
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445257 50 g 50g
11435257 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445257 Supplier Thermo Scientific Alfa Aesar Supplier No. B22644.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90-14-2 | C10H7I | 254.07 g/mol

1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-14-2
Molecular Formula C10H7I
Molecular Weight (g/mol) 254.07
MDL Number MFCD00003876
InChI Key NHPPIJMARIVBGU-UHFFFAOYSA-N
Synonym naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
PubChem CID 7004
IUPAC Name 1-iodonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2I

Specifications

Melting Point 4°C
Density 1.738
Boiling Point 161°C to 162°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.701
Quantity 50 g
Beilstein 1906415
Sensitivity Light Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 254.07
Percent Purity 98%
Chemical Name or Material 1-Iodonaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.