missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane, 98%, stab with copper

Product Code. 11425676
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11425676 100 g 100g
11435676 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11425676

Brand: Thermo Scientific Alfa Aesar B20781.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 628-17-1 | C5H11I | 198.047 g/mol

1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-17-1
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.047
MDL Number MFCD00001100
InChI Key BLXSFCHWMBESKV-UHFFFAOYSA-N
Synonym n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
PubChem CID 12335
IUPAC Name 1-iodopentane
SMILES CCCCCI

Specifications

Melting Point -86°C
Density 1.515
Boiling Point 154°C to 156°C
Flash Point 51°C (123°F)
Odor Characteristic
Linear Formula CH3(CH2)4I
Refractive Index 1.495
Quantity 100 g
UN Number UN1993
Beilstein 1730982
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 198.05
Percent Purity 98%
Chemical Name or Material 1-Iodopentane, stabilized with copper
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodopentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Iodopentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.