missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane, TRC

Product Code. 30398472
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
2.5mL
Product Code. Quantity unitSize
30398472 2.5 mL 2.5mL
1 options
This item is not returnable. View return policy

Product Code. 30398472

Brand: Toronto Research Chemicals TRCI7174532.5ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Iodopentane
CAS 628-17-1
Molecular Formula C5H11I
Quantity 2.5 mL
Synonym Pentane, 1-iodo- (6CI, 8CI, 9CI, ACI), 1-Iodopentane (ACI), 1-Pentyl iodide, 5-Iodopentane, Amyl iodide, n-Amyl iodide, n-Pentyl iodide, NSC 7901, Pentyl iodide
InChI Formula InChI=1S/C5H11I/c1-2-3-4-5-6/h2-5H2,1H3
SMILES CCCCCI
IUPAC Name 1-iodopentane
Molecular Weight (g/mol) 198.04
Formula Weight 197.9905
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.