missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 98+%, stab. with copper

Product Code. 11486987
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486987 500 g 500g
11476987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486987 Supplier Thermo Scientific Alfa Aesar Supplier No. A14388.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-08-4 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
SMILES CC(C)I

Specifications

Melting Point -101°C
Density 1.744
Boiling Point 101°C to 102°C
Flash Point 44°C (111°F)
Assay Percent Range ≥98%, Stabilized with copper
Linear Formula CH3CH2CH2I
Refractive Index 1.505
Quantity 500 g
UN Number UN2392
Beilstein 505937
Sensitivity Light sensitive
Merck Index 14,7862
Solubility Information Fully miscible in water.
IUPAC Name 2-iodopropane
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.