missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 99%, stabilized

Product Code. 10459310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10459310 50 mL Glass bottle 50mL
10409480 250 mL Glass bottle 250mL
10543694 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10459310 Supplier Thermo Scientific Acros Supplier No. 174050500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-08-4 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
IUPAC Name 1-iodopropane
SMILES CC(C)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101.0°C
Color Colorless to Yellow
Density 1.7400g/mL
Boiling Point 101.0°C to 102.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 1.5% max. (Sum of all by-products) (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2I
Refractive Index 1.5030 to 1.5050
Quantity 50 mL
Beilstein 01, 113
Merck Index 15, 7974
Specific Gravity 1.74
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 169.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodopropane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.