missing translation for 'onlineSavingsMsg'
Learn More

1-Iodotetratriacontane, TRC

Product Code. 30401803
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401803 50 mg
30396246 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401803 Supplier Toronto Research Chemicals Supplier No. TRCI72042050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62154-85-2
Molecular Formula C34H69I
Molecular Weight (g/mol) 604.82
IUPAC Name 1-iodotetratriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCI

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C34H69I/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31-32-33-34-35/h2-34H2,1H3
Formula Weight 604.4444
Chemical Name or Material 1-Iodotetratriacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.