missing translation for 'onlineSavingsMsg'
Learn More

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

Product Code. 11976001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976001 5 g 5g
11946001 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976001 Supplier Thermo Scientific Acros Supplier No. 439730050
 Restricted Item
Estimated Shipment: 07-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 827614-66-4 | C13H23BN2O2 | 250.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 827614-66-4
Molecular Formula C13H23BN2O2
Molecular Weight (g/mol) 250.15
MDL Number MFCD05663857
InChI Key YMEBZRNYQBODKB-UHFFFAOYSA-N
Synonym 1-isobutyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutylpyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-1h-pyrazole-4-boronic acid pinacol ester, 1-isobutyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-2-methylpropyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-isobutyl-pyrazole-4-boronic acid pinacol ester, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1h-pyrazole, 1-2-methylpropyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-isobutyl-4-4
PubChem CID 2759392
IUPAC Name 1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0030g/mL
Boiling Point 248°C to 249°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.003
Formula Weight 250.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.