missing translation for 'onlineSavingsMsg'
Learn More

1-Isocyanato-3-(phenoxymethyl)benzene, 97%, Thermo Scientific™

Product Code. 10233723
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10233723 1 g Amber Glass Bottle 1g
10071194 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233723 Supplier Thermo Scientific Maybridge Supplier No. CC63606DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71219-80-2
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.25
MDL Number MFCD09025914
InChI Key CXYJFMVKSSTBDJ-UHFFFAOYSA-N
Synonym 1-isocyanato-3-phenoxymethyl benzene, 3-phenoxymethyl benzenisocyanate, benzene,1-isocyanato-3-phenoxymethyl
PubChem CID 18525913
IUPAC Name 1-isocyanato-3-(phenoxymethyl)benzene
SMILES O=C=NC1=CC=CC(COC2=CC=CC=C2)=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 225.25
Percent Purity 97%
Chemical Name or Material 1-Isocyanato-3-(phenoxymethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.