missing translation for 'onlineSavingsMsg'
Learn More

1-Isoindolinone-5-boronic acid pinacol ester, 96%

Product Code. 15479735
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479735 250 mg 250mg
15418957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479735 Supplier Thermo Scientific Alfa Aesar Supplier No. H52317.MD
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 376584-62-2 | C14H18BNO3 | 259.112 g/mol

1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376584-62-2
Molecular Formula C14H18BNO3
Molecular Weight (g/mol) 259.112
MDL Number MFCD11040393
InChI Key CLACMCLRELMFLJ-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoindolin-1-one, 1-isoindolinone-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-isoindolinone, 1-oxoisoindoline-5-boronic acid pinacol ester, 2,3-dihydro-1h-isoindol-1-one-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroisoindol-1-one, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-isoindol-1-one, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-isoindol-1-one
PubChem CID 21955795
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroisoindol-1-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C(=O)NC3

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 259.11
Percent Purity 96%
Chemical Name or Material 1-Isoindolinone-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.