missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropenyl-2-benzimidazolidinone, 98+%

Product Code. 11348576
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348576 5 g 5g
11338576 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348576 Supplier Thermo Scientific Alfa Aesar Supplier No. L17626.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52099-72-6 | C10H10N2O | 174.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52099-72-6
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00218253
InChI Key XFASJWLBXHWUMW-UHFFFAOYSA-N
Synonym 1-prop-1-en-2-yl-1h-benzo d imidazol-2 3h-one, 1-isopropenyl-2-benzimidazolidinone, 1,3-dihydro-1-1-methylethenyl-2h-benzimidazole-2-one, 1-isopropenylbenzimidazolone, 1,3-dihydro-1-1-methylvinyl-2h-benzimidazol-2-one, 1-prop-1-en-2-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-isopropenyl-1,3-dihydro-benzoimidazol-2-one, 1-isopropenyl-2-benzimidazolinone, 1-isopropenyl-1,3-dihydro-2h-1,3-benzimidazol-2-one, 1-isopropenyl-2,3-dihydro-1h-benzo d imidazol-2-one
PubChem CID 100278
IUPAC Name 3-prop-1-en-2-yl-1H-benzimidazol-2-one
SMILES CC(=C)N1C2=CC=CC=C2NC1=O

Specifications

Melting Point 121°C to 123°C
Quantity 5 g
Beilstein 140587
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 1-Isopropenyl-2-benzimidazolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.